Learn More
Gilac™ Stacking and Nesting Perforated Crate
Description
- Stacking and nesting perforated crate.
- Lightweight.
- Easy to grab hold of thanks to practical, ergonomic integrated handles.
- Stackable up to 2 m.
- 1/2 turn with direction indicator for stacking and nesting.
Specifications
Specifications
| Capacity (English) | 7.4 gal. |
| Capacity (Metric) | 28 L |
| Closure Type | None |
| Color | Blue |
| Material | HDPE |
| Shape | Rectangular |
| Product Type | Crate |
| Stackable | Yes |
| Dimensions (L x W x H) | 600 x 380 x 155 mm |
| Length (Metric) | 600 mm |
| Show More |