Learn More
Stachydrine Chloride, TRC
Specifications
Specifications
| Chemical Name or Material | Stachydrine hydrochloride |
| CAS | 4136-37-2 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C7 H14 N O2 . Cl |
| Quantity | 100 mg |
| Synonym | Pyrrolidinium','2-carboxy-1,1-dimethyl-','chloride (1:1)','(2S)-, Pyrrolidinium','2-carboxy-1,1-dimethyl-','chloride','(2S)- (9CI), Pyrrolidinium','2-carboxy-1,1-dimethyl-','chloride','(S)-, Stachydrine','chloride (6CI,7CI), Stachydrine','hydrochloride (8CI), Cadabine hydrochloride |
| InChI Formula | InChI=1S/C7H13NO2.ClH/c1-8(2)5-3-4-6(8)7(9)10;/h6H,3-5H2,1-2H3;1H/t6-;/m0./s1 |
| SMILES | [Cl-].C[N+]1(C)CCC[C@H]1C(=O)O |
| IUPAC Name | (2S)-1,1-dimethylpyrrolidin-1-ium-2-carboxylic acid;chloride |
| Molecular Weight (g/mol) | 179.64 |
| Show More |