missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Stabilin-1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20663-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes a large, transmembrane receptor protein which may function in angiogenesis, lymphocyte homing, cell adhesion, or receptor scavenging. The protein contains 7 fasciclin, 16 epidermal growth factor (EGF)-like, and 2 laminin-type EGF-like domains as well as a C-type lectin-like hyaluro-binding Link module. The protein is primarily expressed on sinusoidal endothelial cells of liver, spleen, and lymph node.Specifications
Stabilin-1 | |
Polyclonal | |
Unconjugated | |
STAB1 | |
CLEVER 1, FEEL 1, FEEL1, FELE 1, FEX1, KIAA0246, MS 1 antigen, STAB 1, STAB1, Stabilin-1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23166 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.9 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9NY15 | |
STAB1 | |
Peptide | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |