Learn More
Horst Rod sensor, NiCr Ni, 1000°C with plug, DxL=1
Brand:  Horst 70208
Rod sensor, NiCr Ni, 1000°C with plug, DxL=1
Product Code. 10007091
  • ONLINE EXCLUSIVE PRICE
    1900.50 NOK
    1592.77 NOK / Each
    Save 307.73 NOK
    16% Off