Learn More
ST8 alpha-2,8-Sialyltransferase 8B/ST8SIA2 Sheep anti-Human, Polyclonal, R&D Systems™
Sheep Polyclonal Antibody
Brand: R&D Systems AF6590-SP
This item is not returnable.
View return policy
Description
ST8 alpha-2,8-Sialyltransferase 8B/ST8SIA2 Polyclonal specifically detects ST8 alpha-2,8-Sialyltransferase 8B/ST8SIA2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Intracellular Staining by Flow Cytometry, CyTOF-ready.Product Title
Spot an opportunity for improvement?
For Research Use Only