missing translation for 'onlineSavingsMsg'
Learn More

ST7 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 11945-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16889343

  • 3468.15 NOK / 150µL
Estimated Shipment: 19-12-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The gene for this product maps to a region on chromosome 7 identified as an autism-susceptibility locus. Mutation screening of the entire coding region in autistic individuals failed to identify phenotype-specific variants, suggesting that coding mutations for this gene are unlikely to be involved in the etiology of autism. The function of this gene product has not been determined. Transcript variants encoding different isoforms of this protein have been described.
Specifications

Specifications

ST7
Polyclonal
Unconjugated
ST7
ETS7q, FAM4A, FAM4A1, HELG, Protein FAM4A1, Protein HELG, RAY1, SEN4, ST7, TSG7
Rabbit
Antigen Affinity Chromatography
RUO
64213, 7982
-20°C
Liquid
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
0.21 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q99M96, Q9NRC1
St7
ST7 Fusion Protein Ag2547
150 μL
Primary
Mouse, Human
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers