Learn More
ST5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12893-1-AP-20UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16850844
  • 1415.13 NOK / 20µL
Estimated Shipment: 10-10-2023
Description
This gene was identified by its ability to suppress the tumorigenicity of Hela cells in nude mice. The protein encoded by this gene contains a C-terminal region that shares similarity with the Rab 3 family of small GTP binding proteins. This protein preferentially binds to the SH3 domain of c-Abl kinase, and acts as a regulator of MAPK1/ERK2 kinase, which may contribute to its ability to reduce the tumorigenic phenotype in cells. Three alternatively spliced transcript variants of this gene encoding distinct isoforms are identified.
Specifications
Specifications
ST5
Polyclonal
Unconjugated
ST5
DENND2B, HeLa tumor suppression 1, HTS1, p126, ST5
Rabbit
Antigen Affinity Chromatography
RUO
308944, 6764, 76954
-20°C
Liquid
Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P78524, Q924W7
St5
ST5 Fusion Protein Ag3403
20 μL
Primary
Mouse, Human, Rat