missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
TRAP proteins are part of a complex whose function is to bind calcium to the ER membrane and thereby regulate the retention of ER resident proteins. SSR4. also called TRAPD. is assumed to be involved in protein secretion. It is located in the Xq28 region.
Specifications
Specifications
| Antigen | SSR4 |
| Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| Gene | SSR4 |
| Gene Accession No. | P51571 |
| Gene Alias | SSR delta, SSR4, TRAP delta, TRAPD |
| Gene Symbols | SSR4 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?