missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SSR4 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1710.33 NOK - 4836.10 NOK
Specifications
| Antigen | SSR4 |
|---|---|
| Concentration | 0.2 mg/mL |
| Applications | Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16858883
|
Proteintech
11655-2-AP-20UL |
20 μL |
1710.33 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16848883
|
Proteintech
11655-2-AP-150UL |
150 μL |
4836.10 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TRAP proteins are part of a complex whose function is to bind calcium to the ER membrane and thereby regulate the retention of ER resident proteins. SSR4. also called TRAPD. is assumed to be involved in protein secretion. It is located in the Xq28 region.Specifications
| SSR4 | |
| Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P51571 | |
| 6748 | |
| SSR4 Fusion Protein Ag2250 | |
| Primary | |
| -20°C | |
| SSR4 |
| 0.2 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| SSR delta, SSR4, TRAP delta, TRAPD | |
| SSR4 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title