Learn More
SSPN Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26685-1-AP-20UL
This item is not returnable.
View return policy
Description
Component of the dystrophin-glycoprotein complex (DGC), a complex that spans the muscle plasma membrane and forms a link between the F-actin cytoskeleton and the extracellular matrix. Preferentially associates with the sarcoglycan subcomplex of the DGC.Specifications
| SSPN | |
| Polyclonal | |
| Unconjugated | |
| SSPN | |
| DAGA5, Kirsten ras associated protein, KRAG, ospan, NSPN, Sarcospan, SPN1, SPN2, SSPN | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 16651, 500364, 8082 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.67 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q14714, Q62147 | |
| Sspn | |
| SSPN Fusion Protein Ag24511 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |