missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SSEA1 Monoclonal Antibody (MC-480)
Mouse Monoclonal Antibody
1816.00 NOK - 3520.00 NOK
Specifications
| Antigen | SSEA1 |
|---|---|
| Clone | MC-480 |
| Concentration | 1 mg/mL |
| Content And Storage | -20°C |
| Applications | Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
12887742
|
Invitrogen™
MA1022 |
100 μg |
3520.00 NOK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
12807752
|
Invitrogen™
MA1022X |
20 μL |
1816.00 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SSEA1 Monoclonal Antibody for Flow, ICC/IF, IHC (F), IHC (P)
CD15 (Lewis X, Le(x); stage specific embryonic antigen-1, SSEA-1) is a trisacharide determinant (3-fucosyl-N-acetyllactosamine) expressed on several glycolipids, glycoproteins and proteoglycans of various cell types, e.g. granulocytes, mast cells, monocytes, macrophages, cells of gastric mucosa, nervous system or various tumour cells. There are several variants of Lewis x, such as sialyl-Lewis x or sulphated Lewis x. Cells with high surface expression of Le(x) antigen exhibit strong self-aggregation, based on calcium-dependent Le(x)-Le(x) interaction. This process is involved for example in embryo compaction or in autoaggregation of teratocarcinoma cells. Sialyl-Le(x) and its isomer sialyl-Le(a) are ligands of selectins. CD15 expression has been extensively used to confirm diagnosis of Hodgkin's disease.Specifications
| SSEA1 | |
| 1 mg/mL | |
| Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| RUO | |
| PBS with 1mg/mL BSA and 0.05% sodium azide | |
| P22083, Q11127 | |
| 14345, 2526 | |
| BALB/c mouse immunized with F9 teratocarcinoma stem cells (X-irradiated). | |
| Antibody |
| MC-480 | |
| -20°C | |
| Monoclonal | |
| Liquid | |
| IgM | |
| Human, Mouse | |
| FUT4 | |
| FUTIV; Lewis X; SSEA-1 | |
| FUT4 | |
| Primary | |
| Affinity chromatography - MBP |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title