Learn More
SSEA-5 Mouse, anti-Human, Unconjugated, Clone: 8e11, STEMCELL Technologies
Description
- The 8e11 antibody reacts with stage-specific embryonic antigen-5 (SSEA-5), a terminal H type-1 glycan (Fuc1-2Galβ1-3GlcNAcβ1) expressed on the surface of cells in the inner cell mass of the human blastocyst during embryogenesis, and on undifferentiated human embryonic stem (ES) and induced pluripotent stem (iPS) cells
- Expression of SSEA-5 is rapidly down-regulated upon cellular differentiation
- The 8e11 antibody can be used in conjunction with other pluripotency surface markers such as CD9 and CD90, or CD50 and CD200, to sort and remove undifferentiated teratoma-forming cells from incompletely differentiated stem cell cultures
Specifications
Specifications
| Antigen | SSEA-5 |
| Applications | Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Clone | 8e11 |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Quantity | 50 μg |
| Primary or Secondary | Primary |
| Target Species | Human |
| Isotype | IgG1 κ |
| Show More |