missing translation for 'onlineSavingsMsg'
Learn More

SRT 1720 Hydrochloride, MedChemExpress

Product Code. p-200099490
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16488907

Brand: Medchem Express HY15145

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SRT 1720 Hydrochloride is a selective activator of SIRT1 with an EC50 of 0.10 μM, and shows less potent activities on SIRT2 and SIRT3.

SRT 1720 Hydrochloride is a selective activator of SIRT1 with an EC50 of 0.10 μM, and shows less potent activities on SIRT2 and SIRT3.

Specifications

Chemical Name or Material SRT 1720 Hydrochloride
CAS 2060259-60-9
Color Red
Purity Grade Notes Research
Molecular Formula C25H24ClN7OS
Quantity 5 mg
Solubility Information DMSO : 20 mg/mL (39.52 mM; Need ultrasonic) ∣H2O : 15.7 mg/mL (31.03 mM; Need ultrasonic and warming)
SMILES O=C(C1=NC2=CC=CC=C2N=C1)NC3=C(C=CC=C3)C4=CN5C(CN6CCNCC6)=CSC5=N4.Cl
Molecular Weight (g/mol) 506.02
Formula Weight 506.02
Percent Purity 98.21%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.