missing translation for 'onlineSavingsMsg'
Learn More

SRR Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16847974 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16847974 150 μL 150µL
16857974 20 μL 20µL
2 options
This item is not returnable. View return policy

Product Code. 16847974

Brand: Proteintech 179551AP150UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SRR (serine racemase) catalyzes the synthesis of D-serine from L-serine.

Specifications

Antigen SRR
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.19 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SRR
Gene Accession No. Q76EQ0, Q9GZT4, Q9QZX7
Gene Alias ILV1, ISO1, serine racemase, SRR
Gene Symbols Srr
Host Species Rabbit
Immunogen SRR Fusion Protein Ag12449
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27364, 303306, 63826
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.