missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SRR Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17955-1-AP-20UL
This item is not returnable.
View return policy
Description
SRR (serine racemase) catalyzes the synthesis of D-serine from L-serine.Specifications
| SRR | |
| Polyclonal | |
| Unconjugated | |
| SRR | |
| ILV1, ISO1, serine racemase, SRR | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 27364, 303306, 63826 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q76EQ0, Q9GZT4, Q9QZX7 | |
| Srr | |
| SRR Fusion Protein Ag12449 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction