Learn More
SRI-011381 hydrochloride, MedChemExpress
Product Code. 16484276
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16484276 100 mg 100mg
16479526 50 mg 50mg
16475697 5 mg 5mg
16452958 10 mg 10mg
16442556 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 16484276 Supplier Medchem Express Supplier No. HY100347A
MedChemExpress SRI-011381 hydrochloride is an orally active TGF-β signaling agonist, exhibits neuroprotective effects.
SRI-011381 hydrochloride is an orally active TGF-β signaling agonist, exhibits neuroprotective effects.
Specifications
Chemical Name or Material SRI-011381 hydrochloride
Physical Form Solid
CAS 2070014-88-7
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C20H32ClN3O
For Use With (Application) Neuroscience-Neurodegeneration
Solubility Information DMSO : 33.33 mg/mL (91.08 mM; Need ultrasonic) ∣H2O : 25 mg/mL (68.32 mM; Need ultrasonic)
SMILES O=C(NC1CCCCC1)N(CC2=CC=CC=C2)CC3CCNCC3.Cl[H]
Molecular Weight (g/mol) 365.94
Formula Weight 365.94
Percent Purity 99.97%
Grade Research