missing translation for 'onlineSavingsMsg'
Learn More

SRI-011381 hydrochloride, MedChemExpress

Product Code. 16442556
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16442556 25 mg 25mg
16484276 100 mg 100mg
16479526 50 mg 50mg
16475697 5 mg 5mg
16452958 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16442556 Supplier Medchem Express Supplier No. HY100347A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SRI-011381 hydrochloride is an orally active TGF-β signaling agonist, exhibits neuroprotective effects.

SRI-011381 hydrochloride is an orally active TGF-β signaling agonist, exhibits neuroprotective effects.

Specifications

Chemical Name or Material SRI-011381 hydrochloride
Physical Form Solid
CAS 2070014-88-7
Color White
Quantity 25 mg
Purity Grade Notes Research
Molecular Formula C20H32ClN3O
For Use With (Application) Neuroscience-Neurodegeneration
Solubility Information DMSO : 33.33 mg/mL (91.08 mM; Need ultrasonic) ∣H2O : 25 mg/mL (68.32 mM; Need ultrasonic)
SMILES O=C(NC1CCCCC1)N(CC2=CC=CC=C2)CC3CCNCC3.Cl[H]
Molecular Weight (g/mol) 365.94
Formula Weight 365.94
Percent Purity 99.97%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.