missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SRGAP3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1710.33 NOK - 4836.10 NOK
Specifications
| Antigen | SRGAP3 |
|---|---|
| Concentration | 550 μg/mL |
| Applications | Western Blot, Immunofluorescence, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17669676
|
Proteintech
29624-1-AP-20UL |
20 μL |
1710.33 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17659676
|
Proteintech
29624-1-AP-150UL |
150 μL |
4836.10 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SRGAP3 Polyclonal antibody apecifically detetects SRGAP3 antigen in Human, Mouse, Rat Samples. It is validated for Western Blot, Immunofluorescence, ELISA.Specifications
| SRGAP3 | |
| Western Blot, Immunofluorescence, ELISA | |
| Unconjugated | |
| Rabbit | |
| PBS with 0.02% sodium azide and 50% glycerol pH 7.3. | |
| 9901 | |
| SRGAP3 fusion protein Ag31196 | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
| 550 μg/mL | |
| Polyclonal | |
| Liquid | |
| Human, Mouse, Rat | |
| NM_014850 | |
| SRGAP3 | |
| IgG | |
| Antigen affinity purification |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title