Learn More
SREBF1 Mouse anti-Human, Unconjugated, Clone: 1B6G5, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66875-1-IG-20UL
 View more versions of this product
Product Code. 16809145
  • 1710.33 NOK / 20µL
Description
The SREBF1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SREBF1. This antibody recognizes human antigen. The SREBF1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
SREBF1
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Western Blot, Immunohistochemistry, Immunofluorescence
1B6G5
Unconjugated
20 μL
6720
IgG1