Learn More
SR9011, MedChemExpress
Product Code. 16407248
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
2mg
50mg
5mg
Product Code. Quantity unitSize
16407248 10 mg 10mg
16489767 5 mg 5mg
16473257 50 mg 50mg
16417216 100 mg 100mg
16410296 2 mg 2mg
5 options
Product Code. 16407248
Brand: Medchem Express HY16988
MedChemExpress SR9011 is a REV-ERBα/β agonist with IC50s of 790 nM and 560 nM for REV-ERBα and REV-ERBβ, respectively.
SR9011 is a REV-ERBα/β agonist with IC50s of 790 nM and 560 nM for REV-ERBα and REV-ERBβ, respectively.
Specifications
Chemical Name or Material SR9011
Physical Form Solid
CAS 1379686-29-9
Color Light Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C23H31ClN4O3S
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : ≥ 43 mg/mL (89.76 mM)
SMILES O=C(N1CC(CN(CC2=CC=C(Cl)C=C2)CC3=CC=C([N+]([O-])=O)S3)CC1)NCCCCC
Molecular Weight (g/mol) 479.04
Formula Weight 479.04
Percent Purity 98.97%
Grade Research