Learn More
SR1078, MedChemExpress
Product Code. 16486058
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. Quantity unitSize
16486058 10 mg 10mg
16472197 50 mg 50mg
16448667 5 mg 5mg
16406466 100 mg 100mg
4 options
Product Code. 16486058
Brand: Medchem Express HY14422
MedChemExpress SR1078 is a selective agonist of retinoic acid receptor-related orphan receptor α/γ (RORα/RORγ). SR1078 directly binds to the ligand binding domain of RORα and RORγ and increases the transcriptional activity of these receptors, leading to stimulation of RORα/γ target gene transcription.
SR1078 is a selective agonist of retinoic acid receptor-related orphan receptor α/γ (RORα/RORγ). SR1078 directly binds to the ligand binding domain of RORα and RORγ and increases the transcriptional activity of these receptors, leading to stimulation of RORα/γ target gene transcription.
Specifications
Chemical Name or Material SR1078
Physical Form Solid
CAS 1246525-60-9
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C17H10F9NO2
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : 33.33 mg/mL (77.29 mM; Need ultrasonic)
SMILES O=C(NC1=CC=C(C(C(F)(F)F)(C(F)(F)F)O)C=C1)C2=CC=C(C(F)(F)F)C=C2
Molecular Weight (g/mol) 431.25
Formula Weight 431.25
Percent Purity 99.67%
Grade Research