Learn More
Corning™ Square and Rectangular Cover Glasses
GREENER_CHOICE
Product Code. p-4278720 Shop All Corning Products
Change view
Click to view available options
Thickness (Metric):
#1 (0.13 to 0.16 mm)
#1.5 (0.16 to 0.19 mm)
#2 (0.19 to 0.23 mm)
0.17 to 0.25 mm
Length (Metric):
18 mm
22 mm
25 mm
30 mm
40 mm
50 mm
60 mm
Width (Metric):
18 mm
22 mm
24 mm
25 mm
Shape:
Rectangle
Square
Unit Size:
Pack of 1000
Pack of 2000
23 product options available for selection
Product selection table with 23 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Thickness (Metric) Length (Metric) Width (Metric) Shape unitSize
13216778 #1 (0.13 to 0.16 mm) 25 mm 25 mm Square Pack of 2000
13256788 #1.5 (0.16 to 0.19 mm) 50 mm 22 mm Rectangle Pack of 1000
13246778 #1.5 (0.16 to 0.19 mm) 25 mm 25 mm Square Pack of 2000
13286778 #1 (0.13 to 0.16 mm) 30 mm 22 mm Rectangle Pack of 1000
13216788 #1 (0.13 to 0.16 mm) 40 mm 24 mm Rectangle Pack of 1000
13206788 #1 (0.13 to 0.16 mm) 40 mm 22 mm Rectangle Pack of 1000
13258959 #1 (0.13 to 0.16 mm) 50 mm 22 mm Rectangle Pack of 1000
13276778 #2 (0.19 to 0.23 mm) 25 mm 25 mm Square Pack of 1000
13276788 #1.5 (0.16 to 0.19 mm) 40 mm 24 mm Rectangle Pack of 1000
13226788 #1 (0.13 to 0.16 mm) 50 mm 24 mm Rectangle Pack of 1000
13296778 #1 (0.13 to 0.16 mm) 30 mm 24 mm Rectangle Pack of 1000
13286788 #1.5 (0.16 to 0.19 mm) 50 mm 24 mm Rectangle Pack of 1000
13296788 #1.5 (0.16 to 0.19 mm) 60 mm 24 mm Rectangle Pack of 1000
12876333 #1.5 (0.16 to 0.19 mm) 30 mm 22 mm Rectangle Pack of 1000
13236778 #1.5 (0.16 to 0.19 mm) 22 mm 22 mm Square Pack of 2000
13266778 0.17 to 0.25 mm 22 mm 22 mm Square Pack of 1000
13236788 #1 (0.13 to 0.16 mm) 60 mm 24 mm Rectangle Pack of 1000
13256778 0.17 to 0.25 mm 18 mm 18 mm Square Pack of 1000
13266788 #1.5 (0.16 to 0.19 mm) 30 mm 24 mm Rectangle Pack of 1000
13226778 #1.5 (0.16 to 0.19 mm) 18 mm 18 mm Square Pack of 2000
13296768 #1 (0.13 to 0.16 mm) 18 mm 18 mm Square Pack of 2000
13206778 #1 (0.13 to 0.16 mm) 22 mm 22 mm Square Pack of 2000
13246788 #1.5 (0.16 to 0.19 mm) 40 mm 22 mm Rectangle Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 23 options available.
23 options
Product Code. 13216778 Supplier Corning™ Supplier No. 284525
Add to basket
Add to basket
Provides quality, reliability and performance for your laboratory microscopy needs
  • Made from soda-lime glass and borosilicate glass with high clarity
  • Available in square and rectangular shapes, and in three standard sizes and thicknesses
Order Info
Packaged in plastic boxes for protection during storage.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Coverslip
Shape Square
Material Borosilicate Glass
Length (Metric) 25 mm
Width (Metric) 25 mm
Thickness (Metric) #1 (0.13 to 0.16 mm)
Sterility Non-sterile
For Use With (Equipment) Microscopes
Quantity 200/Pk.