Learn More
DWK Life Sciences™ Square Quadrupod, for glass ceramic laboratory protection plate
Product Code. 15262109 Shop All DWK Life Sciences Products
Change view
Click to view available options
Diameter (Metric):
135 mm
155 mm
175 mm
Unit Size:
Pack of 5
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) unitSize
15262109 135 mm Pack of 5
15272109 155 mm Pack of 5
11742909 175 mm Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15262109 Supplier DWK Life Sciences™ Supplier No. 290775302
Stock Up Your Lab
Use promo code 28007
View all promotions
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
Ideal holder for glass ceramic plates.
In the laboratory, solutions are often heated. This step can be carried out using different techniques, e.g. in a water or sand bath, with a heating hood or classically using a Bunsen burner. If the Bunsen burner is used as the heat source, it is recommended to use a tripod or square foot and a corresponding glass ceramic plate. The vessel to be heated is placed on the glass ceramic plate with the Bunsen burner underneath. Contamination of the glass ceramic plate can be removed with the cleaning scraper.
  • holder plate: steel 1.4301, tripod feet: steel 1.4305
  • Made from heat-resistant chrome-nickel steel with
  • Four legs for extra stability
TRUSTED_SUSTAINABILITY
Specifications
Material Chrome-nickel Steel
Diameter (Metric) 135 mm
For Use With (Application) Plate holder for glass ceramic plates; Heating; For Glass Ceramic Laboratory Protection Plate
Height (Metric) 210 mm
No. per Pack 5/Pk.