Learn More
Gilac™ Square Dustbin
Product Code. 17998611
Change view
Click to view available options
Capacity (Metric):
4 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
17998611 4 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17998611 Supplier Gilac™ Supplier No. G038421
  • ONLINE EXCLUSIVE PRICE
    186.90 NOK
    134.54 NOK / Pack of 1
    Save 52.36 NOK
    28% Off
Estimated Shipment: 23-10-2026
Square Dustbin suitable for institutional catering and hotel bedroom use etc.
  • Dustbin suitable for institutional catering and hotel bedroom use etc.
  • With interior white plastic bucket and handle.
  • Highly durable.
  • Stable and easy to maintain.
Specifications
Capacity (Metric) 4 L
Capacity (English) 1.1 gal.
Color White
Shape Square
Autoclavable Not Autoclavable
Material HDPE
Product Type Dust Bin