Learn More
SPTLC1 Mouse anti-Human, Unconjugated, Clone: 2D11D7, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66899-1-IG-20UL
Additional Details : Weight : 0.00010kg
Description
The SPTLC1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SPTLC1. This antibody recognizes human, mouse, rat antigen. The SPTLC1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
SPTLC1 | |
Monoclonal | |
500 μg/mL | |
Mouse | |
Primary | |
Human, Rat, Mouse |
Western Blot, Immunohistochemistry, ELISA | |
2D11D7 | |
Unconjugated | |
20 μL | |
10558 | |
IgG1 |