Learn More
Spreader Blue Polystyrene Wedge-Shaped Spreader
Product Code. 12332048
Change view
Click to view available options
Quantity:
100 bags of 5
25 Bags of 20
Unit Size:
Pack of 500
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12332048 25 Bags of 20 Pack of 500
12342048 100 bags of 5 Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12332048 Supplier Microspec Supplier No. D3
Estimated Shipment: 09-12-2026
Add to basket
Add to basket
High impact wedge-shaped disposable bacterial spreaders
  • High impact polystyrene disposable bacterial spreaders.
  • Wedge shaped to reduce tendency to flip, or traditional “L” shape.
  • Packs of 500 contain re-sealable inner bags of either 20 or 5 sterile spreaders.
  • Manufactured under aseptic conditions
Specifications
Disposable Single-use
Color Blue
Quantity 25 Bags of 20
Shape Wedge-shaped
Material Polystyrene
Product Type Spreader
For Use With (Application) High impact Polystyrene disposable bacterial spreaders