Learn More
MedChemExpress SPQ
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
Specifications
Specifications
| Quantity | 100 mg |
| Storage Requirements | 4°C, protect from light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light) |
| Solubility | H2O : 25 mg/mL (88.86 mM; Need ultrasonic) ∣ DMSO : < 1 mg/mL (insoluble or slightly soluble) |
| Shipping Condition | Room temperature |
| CAS | 83907-40-8 |
| Molecular Weight (g/mol) | 281.33 g/mol |
| Target | HBV |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.