Learn More
Haldenwanger™ Spouted Mortar
Specifications
Specifications
| Capacity (Metric) | 300 mL |
| Diameter (Metric) Outer | 115mm |
| Product Type | Mortar |
| Includes | With spout |
| Type | Mortar |