Learn More
Spirodiclofen, TRC
Specifications
Specifications
| Chemical Name or Material | Spirodiclofen |
| CAS | 148477-71-8 |
| Molecular Formula | C21 H24 Cl2 O4 |
| Quantity | 10 g |
| Synonym | 1-Oxaspiro[4.5]decane','butanoic acid deriv., 3-(2,4-Dichlorophenyl)-2-oxo-1-oxaspiro[4.5]dec-3-en-4-yl 2,2-dimethylbutanoate, BAJ 2740, C21H24Cl2O4, Envidor, Luomanshuangzhi, Spirodiclofen |
| InChI Formula | InChI=1S/C21H24Cl2O4/c1-4-20(2,3)19(25)26-17-16(14-9-8-13(22)12-15(14)23)18(24)27-21(17)10-6-5-7-11-21/h8-9,12H,4-7,10-11H2,1-3H3 |
| SMILES | CCC(C)(C)C(=O)OC1=C(C(=O)OC12CCCCC2)c3ccc(Cl)cc3Cl |
| IUPAC Name | [3-(2,4-dichlorophenyl)-2-oxo-1-oxaspiro[4.5]dec-3-en-4-yl] 2,2-dimethylbutanoate |
| Molecular Weight (g/mol) | 411.32 |
| Formula Weight | 410.1052 |