Learn More
Spirodiclofen 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Product Code. 30321115
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Product Code. 30321115
Brand: Dr. Ehrenstorfer DREA16972950AL100
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Spirodiclofen
CAS 148477-71-8
Analyte or Component Names Spirodiclofen
Shipping Condition +20°C
Molecular Formula C21 H24 Cl2 O4
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
InChI Formula InChI=1S/C21H24Cl2O4/c1-4-20(2,3)19(25)26-17-16(14-9-8-13(22)12-15(14)23)18(24)27-21(17)10-6-5-7-11-21/h8-9,12H,4-7,10-11H2,1-3H3
SMILES CCC(C)(C)C(=O)OC1=C(C(=O)OC12CCCCC2)c3ccc(Cl)cc3Cl
IUPAC Name [3-(2,4-dichlorophenyl)-2-oxo-1-oxaspiro[4.5]dec-3-en-4-yl] 2,2-dimethylbutanoate
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 411.32
Quantity 1 mL
Formula Weight 410.1052 g/mol