Learn More
Buerkle™ Spiral Screw for PowderProof™ Sampler
Product Code. 17627605 Shop All Bürkle Products
Change view
Click to view available options
Length (Metric):
30 cm
90 cm
For Use With (Application):
Granules 2 to 4 mm dia
Granules 2 to 4 mm dia.
Unit Size:
Each
Product Code. Length (Metric) For Use With (Application) unitSize
17627605 90 cm Granules 2 to 4 mm dia. Each
11827383 30 cm Granules 2 to 4 mm dia. Each
11897383 90 cm Granules 2 to 4 mm dia Each
3 options
Product Code. 17627605
Brand: Buerkle™ 53020917
Only null left
Add to basket
Add to basket
Spiral screw is a spare part for use with Buerkle™ PowderProof™ Sampler.
  • Spare part for Buerkle™ PowderProof™ Sampler
TRUSTED_SUSTAINABILITY
Specifications
Product Type Spiral Screw
Material Stainless Steel (V4A)
Autoclavable Autoclavable
For Use With (Application) Granules 2 to 4 mm dia.
Length (Metric) 90 cm