Learn More
Buerkle™ Spiral Screw for PowderProof™ Sampler
Product Code. 11827383 Shop All Bürkle Products
Click to view available options
Length (Metric):
30 cm
90 cm
For Use With (Application):
Granules 2 to 4 mm dia
Granules 2 to 4 mm dia.
Unit Size:
Each
Product Code. 11827383
Brand: Buerkle™ 53020311
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Spiral screw is a spare part for use with Buerkle™ PowderProof™ Sampler.
  • Spare part for Buerkle™ PowderProof™ Sampler
TRUSTED_SUSTAINABILITY
Specifications
Product Type Spiral Screw
Material Stainless Steel (V4A)
For Use With (Application) Granules 2 to 4 mm dia.
Length (Metric) 30 cm