Learn More
Euro Protection™ Spinelle Safety Shoes
Description
- Breathable upper.
- Metal ventilation eyelets on the inner side.
- Padded collar and tongue for snug fit.
- Suede leather and mesh combination upper.
- Breathable 3D mesh lining.
- Removable, textile and foam insole.
- Steel toe cap.
- Stainless steel puncture resistant sole.
- Injected polyurethane midsole.
- Polyurethane outsole.
Specifications
Specifications
| Color | Gray, Orange |
| Liner Material | Breathable 3D mesh lining |
| Material | Leather |
| Size | 43 |
| Sole Features | Removable, textile and foam insole, stainless steel, puncture-resistant sole, injected polyurethan midsole, polyurethane outsole. |
| Sole Material | Polyurethane, Stainless Steel |
| Toe Material | Steel |
| Weight (Metric) | 583 g per shoe in size 8 |
| Certifications/Compliance (Europe) | EN ISO 20345 : 2011 |
| Product Type | Safety Shoe |