missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ SPINeasy™ Plasmid Midiprep Kit

Product Code. 17894126 Shop All MP Biomedicals Products
Click to view available options
Quantity:
25 Preps
Unit Size:
25 preps
This item is not returnable. View return policy

Product Code. 17894126

Brand: MP Biomedicals™ 116539025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Spin column technology for fast and efficient purification of both high and low copy plasmids from bacterial cultures. High-quality plasmid DNA is free of gDNA contamination following a modified alkaline lysis method.

  • SPINeasy Plasmid Midiprep Kit is a silica membrane spin column kit for the purification of high-copy and low-copy number plasmid DNA from 25 to 50 mL of transformed bacterial cultures.
  • Using our specially formulated buffers, bacterial cells are disrupted by alkaline lysis to release plasmid DNA.
  • Cell debris, such as chromosomal DNA and proteins, are precipitated by neutralization and cleared through centrifugation. Subsequently, plasmid DNA in the supernatant is adsorbed on a silica membrane, washed, and eluted in a convenient process using spin column technology.
  • Up to 1 mg of plasmid DNA can be purified through this simple workflow, which is immediately ready for routine molecular biology applications, including cloning, in-vitro transcription, PCR, and restriction digestion.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at Room Temperature, Dry Environment
Isolation Technology Spin Column
Sample Type 25 to 50 mL Bacterial (E. coli) Cultures
Includes 60 mL Resuspension Buffer SD1, 60 mL Alkaline Lysis Buffer SD2, 90 mL Neutralization Buffer SD3, 70 mL Wash Buffer DW1, 12 mL Wash Buffer DW2, 30 mL Elution Buffer SD, 600 µL RNase A, 25 x Column SD with collection tubes
Quantity 25 Preps
compliance-icons
Product Identifier
  • SPINeasy Plasmid Midiprep Kit
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Guanidine Hydrochloride

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.