Learn More
Corning™ Spin-X™ UF Concentrators
GREENER_CHOICE
Product Code. 11323795 Shop All Corning Products
Change view
Click to view available options
MWCO:
10 kDa
100 kDa
30 kDa
5 kDa
50 kDa
Quantity:
12/Pk.
25/Pk.
Volume (Metric):
500 μL
6 mL
20 mL
Filtration Area:
0.5 cm2
2.5 cm2
6 cm2
Unit Size:
Pack of 12
Pack of 25
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. MWCO Quantity Volume (Metric) Filtration Area unitSize
11323795 100 kDa 25/Pk. 500 μL 0.5 cm2 Pack of 25
11383785 5 kDa 25/Pk. 500 μL 0.5 cm2 Pack of 25
11303795 30 kDa 25/Pk. 500 μL 0.5 cm2 Pack of 25
11393785 10 kDa 25/Pk. 500 μL 0.5 cm2 Pack of 25
11313795 50 kDa 25/Pk. 500 μL 0.5 cm2 Pack of 25
11373795 100 kDa 25/Pk. 6 mL 2.5 cm2 Pack of 25
11333795 5 kDa 25/Pk. 6 mL 2.5 cm2 Pack of 25
11343795 10 kDa 25/Pk. 6 mL 2.5 cm2 Pack of 25
11363795 50 kDa 25/Pk. 6 mL 2.5 cm2 Pack of 25
11353795 30 kDa 25/Pk. 6 mL 2.5 cm2 Pack of 25
11383795 5 kDa 12/Pk. 20 mL 6 cm2 Pack of 12
11393795 10 kDa 12/Pk. 20 mL 6 cm2 Pack of 12
11303805 30 kDa 12/Pk. 20 mL 6 cm2 Pack of 12
11323805 100 kDa 12/Pk. 20 mL 6 cm2 Pack of 12
11313805 50 kDa 12/Pk. 20 mL 6 cm2 Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
Product Code. 11323795 Supplier Corning™ Supplier No. 431481
Only null left
Add to basket
Add to basket
Disposable, single use only ultrafiltration devices with polyethersulfone membranes (PES) for the centrifugal concentration and/or purification of biological fluids and aqueous solutions
Spin-X UF centrifugal concentrators offer a simple, one step procedure for concentrating or desalting proteins and other biomolecules with 90% or better recovery.
  • The vertical membrane design and thin channel filtration chamber minimizes membrane fouling and provides fast, high speed concen­trat­ing, even with particle laden solutions.
  • Low binding polyethersulfone (PES) membranes are available with five molecular weight cut-offs (MWCO): 5,000, 10,000, 30,000, 50,000, and 100,000 to meet all of your concentrating needs. Choose an MWCO half to a third smaller than the protein to be concentrated.
  • The MWCO and graduations are printed on the side of the concentrator tube to avoid mix-ups.
  • Spin-X UF 6 and 20 concentrators can be used with either swinging bucket or fixed angle rotors. Spin-X UF 500 con­centrators require fixed angle rotors.
TRUSTED_SUSTAINABILITY
Specifications
Cap Material Polypropylene
Color White
Filter Type Centrifuge Filter
For Use With (Equipment) Fixed Angle Rotor
Material PES
Material (Membrane) PES
MWCO 100 kDa
Filtration Area 0.5 cm2
For Use With (Application) Concentration, desalting of proteins, enzymes, monoclonal antibodies, immunoglobulins, removal of labeled amino acids and nucleotides, HPLC sample preparation, deproteinization of samples, recovery of biomolecules from cell culture supernatants, lysates
Quantity 25/Pk.
Type Ultrafiltration Concentrator
Volume (English) Working 0.016 oz.
Volume (Metric) 500 μL
Volume (Metric) Working 0.5 mL
Height (English) 1.96 in.
Height (Metric) 5 cm
Diameter (English) 0.43 in.
Diameter (Metric) 1.1 cm