Description
- Loads each well of a standard 96-well titer plate with one grinding ball in less than a second.
Specifications
Specifications
| Product Type | Grinding Ball Dispenser |
| For Use With (Equipment) | Grinding Ball and 96-Well Titer Plates of Geno/Grinder |
| Holds | 96-well Microplates |
| Material | Stainless Steel (440 C) |
| Packaging Quantity | 1 Ea. |