Learn More
Cole-Parmer™ XRF Spec-Caps™ Cups Product Code.: 15360542
Cole-Parmer™ XRF Spec-Caps™ Cups
Product Code. p-61604 Shop All SPEX Certiprep Products
33 mm, 8 mm, 100 Pk., Pack of 100
Click to view available options
Diameter (Metric):
30 mm
31 mm
33 mm
38 mm
Height (Metric):
8 mm
9 mm
Quantity:
100 Pk.
1000 Pk.
Unit Size:
Pack of 100
Pack of 1000
Product Code. 15360542
Brand: Cole-Parmer™ 3615
These XRF Spec-Caps™ Cups aid in the production of pressed powder sample disks for XRF, OES, and other analytical techniques.
Aluminum cups aid in the production of pressed-powder sample discs for XRF.
  • Reinforces sample disks to resist chipping and breaking.
  • Makes sample disks easier to remove from the pellet die.
  • Minimizes contamination during sample handling.
  • Protects the bore of the pellet die from abrasive samples.
  • Shallow, thin-walled aluminum cups form the bottom and sides of the finished pellet to help reinforce XRF pellets.
Specifications
For Use With (Equipment) XRF Spectrometers
Type Sample Cup
Description Produces pellets 35 mm x 5 mm
Diameter (English) 1.2 in.
Diameter (Metric) 33 mm
Height (English) 0.31 in.
Height (Metric) 8 mm
Material Aluminum
Quantity 100 Pk.
Product Content Correction
Product Title
Cole-Parmer™ XRF Spec-Caps™ Cups >