Learn More
SPEXQuE™ AOAC Internal Standard Mix, SPEX CertiPrep™
Product Code. 16399987 Shop All SPEX Certiprep Products
Change view
Click to view available options
Quantity:
5 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16399987 5 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16399987 Supplier SPEX Certiprep Supplier No. QUECRAOACIS
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Sensible solutions for pesticide extraction
  • Solvent: Acetonitrile
  • Volume: 5mL
Specifications
Concentration 40 μg/mL
Technique Pesticide Extraction
Type AOAC Internal Standard Mix
Solvent or Matrix Acetonitrile
Quantity 5 mL