Learn More
SPEX CertiPrep™ Epoxy-coated steel Vial Rack
Product Code. 15320492 Shop All SPEX Certiprep Products
Change view
Click to view available options
Material:
Epoxy-coated Steel
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material unitSize
15320492 Epoxy-coated Steel Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15320492 Supplier SPEX CertiPrep™ Supplier No. 6805
This item requires a price update prior to being ordered.
Request a Price for this item here.
Use to hold vials for storage and handling.
  • Durable epoxy-coated steel construction.
  • Holds up to six of the following large grinding vials:6801 Largegrinding vial set 6871 large CR-free grinding vial set
  • Included with the purchase of each 6870L Large Grinding Vial Accessory Package
Specifications
Material Epoxy-coated Steel
For Use With (Application) Holds large grinding vials for storage and handling
For Use With (Equipment) Freezer/Mill™ Cryogenic Grinder
Product Type Vial Rack