Learn More
SPEX CertiPrep™ Accessories for Pipet Washer and Dryer
Product Code. 13206359 Shop All SPEX Certiprep Products
Change view
Click to view available options
Product Type:
Pipet Washer Basin
Pipette Washer Pump, 115V
Pipette Washer Pump, 230V
Unit Size:
1 unit
Product Code. Product Type unitSize
13206359 Pipette Washer Pump, 230V 1 unit
11466068 Pipet Washer Basin 1 unit
15236116 Pipette Washer Pump, 115V 1 unit
3 options
Product Code. 13206359
Brand: SPEX CertiPrep™ PIPPUMP230V
Only null left
Add to basket
Add to basket
Accessories for use with Pipet Washer and Dryer
Pump and Basin both constructed of durable polyethylene.
Certifications
ISO/IEC 17025:2005, ISO 17034:2016, ISO 9001: 2015
Specifications
For Use With Pipet Washers
Height (English) 1 in.
Height (Metric) 3 cm
For Use With (Equipment) For pipet washer and dryer
Product Type Pipette Washer Pump, 230V