Learn More
Thermo Scientific™ SpeedVac™ SRF110 Refrigerated Centrifugal Vacuum Concentrator and Rotors
Product Code. 17364641 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
12-13 mm Rotor
12-17 mm Rotor
28 mm Rotor
Hexagonal Microcentrifuge Rotor
Microplate Rotor
Vacuum Concentrator
Vacuum Concentrator Kit
Vacuum Concentrator Universal Vacuum Kit
Electrical Requirements:
230 V, 50 Hz, 8 A
N/A
Includes:
Concentrator
Oil Pump, Vapor Trap, Accessories
Rotor
Universal Vacuum System, Rotor, Accessories
Unit Size:
Each
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Electrical Requirements Includes unitSize
17364641 12-13 mm Rotor N/A Rotor Each
17374641 Hexagonal Microcentrifuge Rotor N/A Rotor Each
17384641 28 mm Rotor N/A Rotor Each
17394641 12-17 mm Rotor N/A Rotor Each
17304651 Microplate Rotor N/A Rotor Each
17314651 Vacuum Concentrator 230 V, 50 Hz, 8 A Concentrator Each
17324651 Vacuum Concentrator Kit 230 V, 50 Hz, 8 A Oil Pump, Vapor Trap, Accessories Each
17334651 Vacuum Concentrator Universal Vacuum Kit 230 V, 50 Hz, 8 A Universal Vacuum System, Rotor, Accessories Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 17364641 Supplier Thermo Scientific™ Supplier No. RF108
  • ONLINE EXCLUSIVE PRICE
    15415.00 NOK
    14623.47 NOK / Pack of 1
    Save 791.53 NOK
    5% Off
In Stock
Add to basket
Add to basket
Process heat sensitive biological samples with confidence using SRF110. This Refrigerated SpeedVac system uses state-of-the-art centrifugation, vacuum, and heat technologies for removing solvents and concentrating heat sensitive biological samples while maintaining sample integrity.
SpeedVac™ SRF110 modular refrigerated vacuum concentrator provides flexibility to your lab. It offers unique programming capabilities to maintain chamber temperature pre and post sample run to prevent thermal degradation of heat sensitive samples such as proteins, enzymes, and other biomolecules. They are ideal for evaporation of aqueous based solvents, buffers, and HPLC fractions. For a convenient, one model shopping experience select from SRF110P1 (modular configuration) and SRF110P2 (Semi integrated configuration) kit options. Each kit is designed to include components required for convenient SpeedVac setup.
SRF110 modular refrigerated vacuum concentrator provides flexibility to any benchtop space.  SRF110 can be configured depending on your applications need. Select from wide range of suitable ThermoFisher Scientific components such as pumps, Vapor Trap, and additional accessories to configure your custom vacuum concentration setup.  For a convenient, one model shopping experience select from SRF110P1 (modular configuration) and SRF110P2 (Semi integrated configuration) kit options. Each kit is designed to include components required for convenient SpeedVac setup.
  • SRF110 offers unique programming capability to maintain chamber temperature pre and post sample run to prevent thermal degradation of heat sensitive samples such as proteins, enzymes, and other biomolecules
  • Ideal for evaporation of non-aggressive, aqueous based solvents, buffers, HPLC fractions, etc.
  • 3 Preset program with option to store up to 9 programs
  • Temperature range -4°C up to 100°C (1°C increment)
  • Self aligning and quick release rotor mechanism
  • Hexagonal rotor (RF132) included with purchase of SRF110
  • Refer to list of rotors available for purchase and to be used with SRF110 model
  • Electricals- 115V AC (60Hz, 16Amps) and 230V AC (50Hz, 8Amps)
  • Certifications- UL Certified, CE, UKCA, KC, RCM Marked.
  • F Gas Compliance- Yes (EPA-SNAP and EU)
  • SRF110P1 kit includes Refrigerated SpeedVac (SRF110), Hexagonal rotor (RF132), an Oil pump (VLP120-115), a vapor Trap (RVT450-115), Universal Tubing (UTP-TYG), 4L capacity Glass Condensation Flask (GCF400), Glass Cap (FC400), and 1L Bottle of heat transfer fluid Cryocool™ (SCC1)
  • SRF110P2 kit includes SRF110P2 Kit includes Refrigerated SpeedVac (SRF110), Hexagonal rotor (RF132), Universal Vacuum System (UVS450), Universal Tubing (UTP-TYG), 4L capacity Glass Condensation Flask (GCF400), Glass Cap (FC400), and 1L Bottle of heat transfer fluid Cryocool™ (SCC1)
Warranty
Out of Box
TRUSTED_SUSTAINABILITY
Specifications
Certifications None
Holds 40 x 1.5, 2.0 mL; 16 x 3 mL
Includes Rotor
Type 12-13 mm Rotor
Description 12-13 mm Rotor
Electrical Requirements N/A
Material Aluminum
For Use With (Equipment) SRF110