Learn More
3M™ Speedglas™ Outer Protection Plate 9000
Product Code. 15729637
Change view
Click to view available options
Style:
Heat Resistant
Scratch Resistant
Standard
Unit Size:
Pack of 10
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Style unitSize
15729637 Heat Resistant Pack of 10
15780048 Standard Pack of 10
15799627 Scratch Resistant Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15729637 Supplier 3M™ Supplier No. 7000000206
  • ONLINE EXCLUSIVE PRICE
    969.00 NOK
    806.93 NOK / Pack of 10
    Save 162.07 NOK
    16% Off
Estimated Shipment: 19-11-2026
Add to basket
Add to basket
3M™ Speedglas™ Outer Protection Plate 9000 for use with 3M™ Speedglas™ Welding Helmet Series 9000.
  • Replacement spare part to the original protection plate if any pits or scratches occur
  • For maximum safety and comfort, make it a habit to check your equipment regularly
  • Regularly clean with a soft cloth and replace if any pits or scratches occur
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EN 166‎, EN166:BT
For Use With (Equipment) Speedglas Welding Helmet Series 9000
Lens Tint Clear
Product Type Helmet Outside Protection Plate
Style Heat Resistant