Learn More
3M™ Speedglas™ Neck Protection
Product Code. 15829634 Shop All 3M Products
Change view
Click to view available options
For Use With (Equipment):
Speedglas Welding Helmet Series 100, 9000, SL and 9100 MP
Speedglas Welding Helmet Series 9100
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15829634 Speedglas Welding Helmet Series 9100 Each
15799757 Speedglas Welding Helmet Series 100, 9000, SL and 9100 MP Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15829634 Supplier 3M™ Supplier No. 7000000230
  • ONLINE EXCLUSIVE PRICE
    935.00 NOK
    677.76 NOK / Pack of 1
    Save 257.24 NOK
    27% Off
Estimated Shipment: 24-02-2027
3M™ Speedglas™ Neck Protection offers extended protection for Speedglas™ Welding Helmet.
  • Made in flame-retardant textiles for protection against sparks and flames
  • For use with the complete helmet assembly to help protect the user from welding spark, spatter, and radiation on the ears and neck
TRUSTED_SUSTAINABILITY
Specifications
Color Black
For Use With (Equipment) Speedglas Welding Helmet Series 9100
Product Type Neck Protection