Learn More
3M™ Speedglas™ Inner Cover Plate 9002X
Click to view available options
For Use With (Application):
Increase 1 shade
Increase 2 shades
Protection
Unit Size:
Pack of 5
Description
- The inner cover plate is designed to protect the inside surface of the filter from spark and spatter
- Replace anytime the plate becomes deeply pitted, scratched or too dirty to clean with soft cloth or lens paper
- Regularly clean with a soft cloth and replace if any pits or scratches occur
- Can be used to increase +1 or +2 dark shades of the welding filter
- Failure to use an inside protection plate with the filter voids the warranty if spark or spatter cause damage to the glass
Specifications
Specifications
| Color | Clear |
| Dimensions (L x W) | 53 x 103 cm |
| For Use With (Equipment) | 3M Speedglas Welding Filter |
| For Use With (Application) | Increase 2 shades |
| Product Type | Inner Cover Plate |