Learn More
Spectromelt™ C 10, Supelco™
Product Code. 15853911
Change view
Click to view available options
Weight (Metric):
5 kg
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Weight (Metric) Unit Size
15853911 5 kg 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15853911 Supplier Supelco Supplier No. 1139905000
 Restricted Item
Estimated Shipment: 13-11-2026
For X-ray fluorescence analysis
Grinding and tabletting aid for X-ray fluorescence spectroscopy
TRUSTED_SUSTAINABILITY
Specifications
Weight (Metric) 5 kg