Learn More
Spectromelt™ A 14, Supelco™
Product Code. 15857870
Change view
Click to view available options
Weight (Metric):
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Weight (Metric) Unit Size
15857870 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15857870 Supplier Supelco Supplier No. 1117241000
 Restricted Item
Estimated Shipment: 13-11-2026
51% Di-lithium tetraborate/ 27% lanthane(III) oxide/ 12% lithium metaborate/ 10% lithium fluoride
TRUSTED_SUSTAINABILITY
Specifications
Weight (Metric) 1 kg