Learn More
Spectromelt™ A 12, Supelco™
Product Code. 15845610
Change view
Click to view available options
Weight (Metric):
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Weight (Metric) Unit Size
15845610 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15845610 Supplier Supelco Supplier No. 1118021000
 Restricted Item
Estimated Shipment: 06-01-2027
66% Di-lithium tetraborate/34% lithium metaborate, for X-ray fluorescence analysis
TRUSTED_SUSTAINABILITY
Specifications
Weight (Metric) 1 kg