Description
- Designed for imaging-based workflows in microplate-based assays
- Supports visualisation and analysis of samples in microplate formats
- Enables image capture to complement absorbance and fluorescence assay workflows
- Supports cell-based assay applications requiring imaging analysis
- Provides integrated workflow support with compatible SpectraMax systems
- Designed to support microplate-based imaging workflows across life science applications
- Integrated with SoftMax Pro Software for data acquisition and analysis
Specifications
Specifications
| Product Type | Imaging Cytometer |
| For Use With (Equipment) | Microplate Reader |
| Sterility | Non-sterile |