Learn More
Labnet™ Spectrafuge™ 6C Centrifuge
Product Code. 11441997
Change view
Click to view available options
Max. RCF:
4,000 x g
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Max. RCF Unit Size
11441997 4,000 x g Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11441997 Supplier Labnet™ Supplier No. C0060230V
  • ONLINE EXCLUSIVE PRICE
    12894.00 NOK
    10595.61 NOK / Pack of 1
    Save 2298.39 NOK
    17% Off
 Restricted Item
In Stock
Rugged construction and a small footprint means the Spectrafuge™ 6C Compact Research Centrigufe is ready to provide years of trouble-free operation.
  • The Spectrafuge 6C has been designed for quick production of PPP (platelet poor plasma) and PRP (platelet rich plasma) as well as other applications in the clinical or research laboratory.
  • The removable fixed angle rotor of the Spectrafuge 6C accepts 6 x 15 mL tubes, and with the adapers will accommodate all common blood draw tubes.
  • The variable speed allows for a variety of applications to be performed. A timer controls operation from 30 seconds to 30 minutes. The time can also be set to continuous, by selecting the 'On' position, or momentary, by pressing and holding the start knob.
  • The control panel of the centrifuge feature easy to turn knobs and large digital displays for time and speed.
  • During operation of the centrifuge, the lid is locked. Once the rotor has come to a stop, both the time and speed displays flash '00' to signify that the run has been completed. The lid can now be opened by pressing the lid knob.
  • A small footprint allows the Spectrafuge 6C to fit on even the most crowded bench.
  • Its rugged construction will provide years of trouble free operation.
  • The centrifuge can be operated on the bench or in a cold room.
Specifications
Max. Speed 6,500 rpm
Max. RCF 4,000 x g
Timer 0.5 to 30 min. or Continuous
Dimensions (HWD) 24 x 21 x 18 cm (9.5 x 8.25 x 7 in.)
Net Weight 4.5 kg (10 lb.)
Frequency 50/60Hz
Voltage 230 V
Speed Control Digital - Variable (100 rpm inc.)