Learn More
Spectrum Labs™ Spectra/Chrom™ Aqueous Series Columns
Click to view available options
Length (Metric):
300 mm
600 mm
Diameter (Metric) Outer:
25 mm
50 mm
Unit Size:
Each
Specifications
Specifications
| For Use With (Equipment) | For use with standard buffers |
| Quantity | 1 Each |
| Diameter (Metric) Inner | 50 mm |
| Type | Aqueous Column |
| Length (Metric) | 600 mm |
| Material | Polypropylene |
| Diameter (Metric) Outer | 50 mm |